Zwitterionic columns, CHIRALPAK® ZWIX
Chiralpak® ZWIX(-), Semi-preparatief, 5 µm, 10,00 mm, 250 mm
Catalogus # 554-4595
Leverancier: CHIRAL TECHNOLOGIES
Ga naar:
Product Details & Documents
The CHIRALPAK® ZWIX columns were developed for separation of a great variety of zwitterionic molecules, specifically, for separation of underivatised amino acids.
ZWIX selectors are zwitterionic molecules that incorporate both anion- and cation-exchange functional groups. These ZWIX selectors exhibit exceptional enantioseparation capabilities toward zwitterionic molecules such as amino acids and small peptides.
Specificaties
- Kolomtype:Semi-preparatief
- Ø inw.:10,00 mm
- Partikelgrootte:5 µm
- Omschrijving:Chiralpak® ZWIX(-)
- Lengte:250 mm
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



