Orders LinkContactUsLinkComponent
Corning® Trypsin
Catalogus # 25-053-CI
Leverancier:   Corning
Corning® Trypsin
Catalogus # 25-053-CI
Leverancier:  Corning
CAS-nummer:  
Corning® Trypsin
Corning® Trypsin
CAS-nummer:  
Price
115,00 €
Log in voor beschikbaarheid
1 * 6 ST
Product Details & Documents
0,05% Trypsin/0,53 mM EDTA in HBSS (Hank’s Balanced Salt Solution) with sodium bicarbonate, without calcium and magnesium.
  • Trypsin EDTA 1X 0,05% Trypsin/0,53 mM EDTA in HBSS with sodium bicarbonate, without calcium and magnesium
  • Porcine Parvovirus tested
Trypsin is used to enzymatically release adherent cells from tissue culture plates for passaging. Divalent cations such as calcium and magnesium, which are often present in the cell culture environment, inhibit this action. EDTA sequesters these ions and thus enhances the efficacy of trypsin. This product contains trypsin at a concentration of 0,05% with 0,53 mM EDTA, and does not contain sodium bicarbonate.
Specificaties
Specificaties
  • Cat. No.:
    25-053-CI
  • Enzym naam:
    Trypsin
  • CAS No.:
    9002-07-7
Specificaties