Orders LinkContactUsLinkComponent
Jars with screw cap
Jars with screw cap
with polypropylene screw cap, irradiated, 150 ml
Catalog # 216-2029
Supplier:   Anicrin
Jars with screw cap
Catalog # 216-2029
Supplier:  Anicrin
Jars with screw cap
Jars with screw cap
with polypropylene screw cap, irradiated, 150 ml
Price
114,00 €
Log in for availability
1 * 250 ST
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
  • Twist cap for quick opening and closing
  • Very good temperature resistance
  • Very good chemical resistance
Caution: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Specifications
  • Type:
    with polypropylene screw cap, irradiated
  • Capaciteit:
    150 ml
  • Seq:
    03