Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Guanidiniumchloride

Guanidiniumchloride

Plastic bucket, 25 kg
Catalogus # A1499.9025
Irriterend
Leverancier:   PanReac AppliChem
Guanidiniumchloride
Catalogus # A1499.9025
Leverancier:  PanReac AppliChem
CAS-nummer:  50-01-1
Guanidiniumchloride
Guanidiniumchloride

Plastic bucket, 25 kg

CAS-nummer:  50-01-1
Price
2 850,00 €
Log in voor beschikbaarheid
1 * 25 kg
Product Details & Documents
Synoniemen:  Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
  • CAS-nummer:50-01-1
  • Formule:NH₂C(=NH)NH₂·HCl
  • MDL-nummer:MFCD00013026
  • Molecular Weight:95.53 g/mol
  • Merck index:13,04578
  • Smelten Pt:185 °C
  • Dichtheid:1,354 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:200-002-3
Specificaties

Specificaties

  • Verpakking:
    Plastic bucket
  • VPE:
    25 kg

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.