Orders LinkContactUsLinkComponent
Staining dishes
Staining dishes
Staining dish with seperate lid
Catalogus # BOPT10-33
Leverancier:   BIO OPTICA
Staining dishes
Catalogus # BOPT10-33
Leverancier:  BIO OPTICA
Staining dishes
Staining dishes
Staining dish with seperate lid
Price
321,00 €
Log in voor beschikbaarheid
1 * 12 ST
Product Details & Documents
These staining dishes are made of PET (polyethylene terephthalate) stiffened with glass fibres. They are resistant against solvents especially in cytology and histology staining processes.
  • Resist temperature between 0 to 170 °C
  • Can be used with immunohistochemistry procedures
Specificaties
  • Omschrijving:
    Staining dish with seperate lid
  • Seq:
    02
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Rack for 25 slides with plastic handle
    Rack for 25 slides with plastic handle
    BOPT10-42
    Log in voor beschikbaarheid
    1 * 6 ST - 96,00 €