Caps, closed top
With bonded PTFE Liner, bulk, 33 - 400 mm
Catalogus # EPSCB167-3300BL
Leverancier: EP SCIENTIFIC
Caps, closed top
Catalogus # EPSCB167-3300BL
Leverancier: EP SCIENTIFIC
Ga naar:
Product Details & Documents
PP, white.
- Very good chemical resistance
- Very good temperature resistance
Unprocessed caps are ideal for environmental sample bottles, jars, jugs and vials.
Certificeringen: Manufactured under an ISO certified quality management system
Specificaties
- Sluiting diam:33 - 400 mm
- Omschrijving:With bonded PTFE Liner, bulk
- Seq:02
- Cat. No.:EPSCB167-3300BL
- Milieu certificering:NAR Certificate of Retirement
- Environmentally Preferable:Lage impact op de fabricage