Disposable filters, Optimizer®
D disposable filter with 6,35 mm connection, 6,35 mm vent/drain compression seal fittings, Ø×L: 69×113 mm, 0,05 µm, 1800 cm²
Catalogus # ENTECWSF0S2S3M
Leverancier: ENTEGRIS
Ga naar:
Product Details & Documents
Photochemical filter with superior contaminant retention and low hold-up volume performance in sub-10 to 180 nm technology nodes.
Specificaties
- Filtreeroppervlakte:1800 cm²
- Poriëngrootte:0,05 µm
- Type:D disposable filter with 6,35 mm connection, 6,35 mm vent/drain compression seal fittings, Ø×L: 69×113 mm
- Seq:09
- Cat. No.:ENTECWSF0S2S3M