Orders LinkContactUsLinkComponent
Conductivity standards
Conductivity standards
Sodium chloride standard solution, pour-top, 491 mg/l NaCl, 1000 µS/cm, 50 mL
Catalogus # HACH1440026
Leverancier:   Hach
Conductivity standards
Catalogus # HACH1440026
Leverancier:  Hach
CAS-nummer:  
Conductivity standards
Conductivity standards
Sodium chloride standard solution, pour-top, 491 mg/l NaCl, 1000 µS/cm, 50 mL
CAS-nummer:  
Price
16,20 €
Log in voor beschikbaarheid
1 * 50 mL
Product Details & Documents
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.
Specificaties
Specificaties
  • VPE:
    50 mL
  • Omschrijving:
    Sodium chloride standard solution, pour-top, 491 mg/l NaCl
  • Geleidbaarheid:
    1000 µS/cm