Orders LinkContactUsLinkComponent
Accessories for pH electrodes
Accessories for pH electrodes
pH electrode storage solution, 250 ml
N° de catalogue JENW99959-60
Fournisseur:   Cole-Parmer
Accessories for pH electrodes
N° de catalogue JENW99959-60
Fournisseur:  Cole-Parmer
Accessories for pH electrodes
Accessories for pH electrodes
pH electrode storage solution, 250 ml
Prix
57,00 €
Connexion Pour la disponibilité
1 * 100 mL
Product Details & Documents
Services: Installatie, IQOQ, onderhoud of reparatie nodig? Mail naar [email protected]
Spécifications
  • Omschrijving:
    pH electrode storage solution, 250 ml