0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
MOBCAP COLOR: YELLOW SIZE: 52CM DESCRIPTION: DETECTABLE WITH DOUBLE LATEXFREE ELASTIC BAND MATERIAL: POLYPROPYLENE THICKNESS: 12-14 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 ST
GUNA140016
:
VWR Collection
MOBCAP COLOR: YELLOW SIZE: 52CM DESCRIPTION: DETECTABLE WITH DOUBLE LATEXFREE ELASTIC BAND MATERIAL: POLYPROPYLENE THICKNESS: 12-14 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 ST
GUNA140016
:
VWR Collection
Frequently Bought Together
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
VWR®, gezichtstissues
: VWR Collection
VWR®, Nitrilhandschoenen
: VWR Collection
pH-teststrips, MQuant®, Supelco®
: Merck
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Sealing film for manual application, Parafilm® M
: HEATHROW
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Nitril handschoenen, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR®, Serologische pipetten, standaardreeks
: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschoenen
: VWR Collection
VWR®, Transferpipetten, Hoge Prestaties
: VWR Collection
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals