Sondes
Interne vervangende temperatuursonde voor EBI 300 TH, TPH 400, TPH 300 (NTC, capacitive), –30… +70 °C; 0 - 100 % RH, RH: ±3% between 10 - 90% (at +25 °C); ±5 % for the remaining measuring range
T: ±0,5 °C (+20… +40 °C); ±1 °C for the remaining measuring range
T: ±0,5 °C (+20… +40 °C); ±1 °C for the remaining measuring range
Catalogus # 620-2262
Leverancier: EBRO
Ga naar:
Product Details & Documents
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature.
: Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
Specificaties
- Meetbereik:–30… +70 °C; 0 - 100 % RH
- Omschrijving:Interne vervangende temperatuursonde voor EBI 300 TH, TPH 400
- Juistheid:RH: ±3% between 10 - 90% (at +25 °C); ±5 % for the remaining measuring range
T: ±0,5 °C (+20… +40 °C); ±1 °C for the remaining measuring range - Type:TPH 300 (NTC, capacitive)