Sondes
External replacement probe for EBI 310TX- P4, Ø×L: 5×45 mm, blunt, with 3 m metal wrapped cable, 100...400 °C, —
Catalogus # 620-2765
Leverancier: EBRO
Ga naar:
Product Details & Documents
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature.
: Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
Specificaties
- Meetbereik:100...400 °C
- Omschrijving:External replacement probe for EBI 310TX- P4, Ø×L: 5×45 mm, blunt, with 3 m metal wrapped cable
- Seq:08
- Juistheid:—