Sondes
External replacement probe for EBI 310TX- P1, Ø×L: 5×45 mm, blunt, with 3 m PTFE cable, −200...+200 °C, —
Catalogus # 620-2762
Leverancier: EBRO
Ga naar:
Product Details & Documents
Use the humidity/temperature probes (with an appropriate measuring instrument) for the reliable measurement of relative humidity and temperature.
Diensten: Voor installatie, IQOQ, erkende kalibratie onder ISO17025, onderhoud en reparatie mail naar [email protected]
Documents
Specificaties
- Meetbereik:−200...+200 °C
- Omschrijving:External replacement probe for EBI 310TX- P1, Ø×L: 5×45 mm, blunt, with 3 m PTFE cable
- Seq:05
- Juistheid:—