Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
VWR®, DNA ladders
VWR®, DNA ladders
PCR-ladder, 100 tot 3000 bp, voldoende voor 250 banen (lading 2 μl per gel), 1×0,5 ml, 1 KIT
Catalog # 733-2579
Supplier:   VWR Chemicals
VWR®, DNA ladders
Catalog # 733-2579
Supplier:  VWR Chemicals
CAS Number:  
VWR®, DNA ladders
VWR®, DNA ladders
PCR-ladder, 100 tot 3000 bp, voldoende voor 250 banen (lading 2 μl per gel), 1×0,5 ml, 1 KIT
CAS Number:  
Price
214,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
VWR® DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.
  • Supplied in loading buffer, ready to use directly on the gel
  • Mass-calibrated bands for DNA quantification
  • PCR DNA ladder has a 1000 bp band that is extra bright to serve as reference point
High range DNA ladder molecular range: 200 to 12000 bp; mass-calibrated bands from 6 to 40 ng/band for DNA quantification.
Low range DNA ladder molecular range: 100 to 1000 bp; mass-calibrated bands from 8 to 40 ng/band for DNA quantification.
PCR DNA ladder molecular range: 100 to 3000 bp; mass-calibrated bands from 10 to 30 ng/band for DNA quantification.
Delivery information: Each DNA ladder is supplied in 0,5 ml packs sufficient for 250 lanes.
Specifications
Specifications
  • VPE:
    1 KIT
  • Omschrijving:
    PCR-ladder, 100 tot 3000 bp, voldoende voor 250 banen (lading 2 μl per gel), 1×0,5 ml
Specifications