VWR®, DNA ladders
PCR-ladder, 100 tot 3000 bp, voldoende voor 250 banen (lading 2 μl per gel), 1×0,5 ml, 1 KIT
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
VWR® DNA ladders are supplied in a loading buffer that is ready to use on agarose and polyacrylamide DNA gels. The ladders are suitable with TBE, TAE, SB and LB electrophoresis systems.
- Supplied in loading buffer, ready to use directly on the gel
- Mass-calibrated bands for DNA quantification
- PCR DNA ladder has a 1000 bp band that is extra bright to serve as reference point
High range DNA ladder molecular range: 200 to 12000 bp; mass-calibrated bands from 6 to 40 ng/band for DNA quantification.
Low range DNA ladder molecular range: 100 to 1000 bp; mass-calibrated bands from 8 to 40 ng/band for DNA quantification.
PCR DNA ladder molecular range: 100 to 3000 bp; mass-calibrated bands from 10 to 30 ng/band for DNA quantification.
Delivery information: Each DNA ladder is supplied in 0,5 ml packs sufficient for 250 lanes.
Documents
Specifications
Specifications
- VPE:1 KIT
- Omschrijving:PCR-ladder, 100 tot 3000 bp, voldoende voor 250 banen (lading 2 μl per gel), 1×0,5 ml