Natriumhydroxide solution 50%, Biotech Reagent, J.T.Baker®
1.000 L
: Avantor
: Caustic soda
- :1310-73-2
- :NaOH
- :MFCD00003548
- :40 g/mol
- :1823
- :8,II
- Assay (NaOH):49 - 51 %
- Appearance (clear, colorless liquid):passes test
- Potassium (K):max. 0.01 %
- Identification (Na):passes test
- Sodium Carbonate (Na2CO3):max. 1.0 %
- Trace Impurities (in ppm) Chlorides (as Cl):max. 5
- Trace Impurities (in ppm) Iron (Fe):max. 2
- Trace Impurities (in ppm) Heavy Metals (as Pb):max. 1
- VPE:1.000 L



