Natriumhydroxide solution 50%, Biotech Reagent, J.T.Baker®
5 L
Leverancier: Avantor
Natriumhydroxide solution 50%, Biotech Reagent, J.T.Baker®
Catalogus # 2622-35
Leverancier: Avantor
CAS-nummer: 1310-73-2
Ga naar:
Product Details & Documents
Synoniemen: Caustic soda
- CAS-nummer:1310-73-2
- Formule:NaOH
- MDL-nummer:MFCD00003548
- Molecular Weight:40 g/mol
- UN:1823
- ADR:8,II
Documents
Specificaties
Specificatie testresultaten
- Assay (NaOH):49 - 51 %
- Appearance (clear, colorless liquid):passes test
- Potassium (K):max. 0.01 %
- Identification (Na):passes test
- Sodium Carbonate (Na2CO3):max. 1.0 %
- Trace Impurities (in ppm) Chlorides (as Cl):max. 5
- Trace Impurities (in ppm) Iron (Fe):max. 2
- Trace Impurities (in ppm) Heavy Metals (as Pb):max. 1
Specificaties
- VPE:5 L



