Anti-EPEC Rabbit Polyclonal Antibody
5 µg
Catalogus # BIRBORB13394-5
Marketsource
Leverancier: Biorbyt
Ga naar:
Product Details & Documents
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)
Type: Primary
Antigen: EPEC
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Bacteria
Specificaties
- Maat:5 µg
- Western blot:Yes
- Clonality:Polyklonaal
- antigeen symbool:EPEC
- Purification:purified by affinity chromatography
- Type antilichaam:Primair
- Isotype:IgG
- Host:Rabbit
- Opslagbuffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Shipping temperature:Ice
- ImmunoChemie:Yes
- Cat. No.:BIRBORB13394-5
- Conjugatie:Onconjugeerd
- Reactivity:Bacteria
- Opslagtemperatuur:Cool
- Formulier:liquid
- Immunogeen:Synthetic peptide



