Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
QuEChERS extraction kits

QuEChERS extraction kits

Non-buffered QuEChERS extraction tubes with ceramic homogenisers, Original QuEChERS method, with 6 g magnesium sulphate and 1,5 g sodium chloride, for analysing pesticide residues in fruits and vegetables (15 g samples)
Catalogus # HEWL5982-5555CH
Leverancier:   Agilent
QuEChERS extraction kits
Catalogus # HEWL5982-5555CH
Leverancier:  Agilent
QuEChERS extraction kits
QuEChERS extraction kits

Non-buffered QuEChERS extraction tubes with ceramic homogenisers, Original QuEChERS method, with 6 g magnesium sulphate and 1,5 g sodium chloride, for analysing pesticide residues in fruits and vegetables (15 g samples)

Price
224,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Step 1: Extraction
Choose the extraction salt packet based on your method of analysis, AOAC or EN. The buffered extraction salts are amenable for more liable pesticides. Adding solvent and then salts to a comminuted fruit or vegetable sample (10 g or 15 g) enables you to extract the pesticides of interest into the organic layer. However, adding a food sample with a high percentage of water directly to the salts may create an exothermic reaction that can affect your analyte recoveries.

  • Available with or without 50 ml centrifuge tubes and caps
  • Include MgSO₄, NaCl, or other salts for buffering; pre-weighed in anhydrous packet

QuEChERS salts and buffers are packaged in anhydrous packages. This allows you to add them after adding your solvent to the sample, as specified in QuEChERS methodologies.

Specificaties
  • Omschrijving:
    Non-buffered QuEChERS extraction tubes with ceramic homogenisers, Original QuEChERS method, with 6 g magnesium sulphate and 1,5 g sodium chloride, for analysing pesticide residues in fruits and vegetables (15 g samples)
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.