Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
QuEChERS dispersive kits
QuEChERS dispersive kits
Tube with 400 mg primary secondary amine, 400 mg GCB, 1200 mg magnesium sulphate: with ceramic homogenisers, removes polar organic acids, some sugars and lipids, and carotinoides and chlorophyll, not for use with planar pesticide, 15 ml
Catalogus # HEWL5982-5258CH
Leverancier:   Agilent
QuEChERS dispersive kits
Catalogus # HEWL5982-5258CH
Leverancier:  Agilent
QuEChERS dispersive kits
QuEChERS dispersive kits
Tube with 400 mg primary secondary amine, 400 mg GCB, 1200 mg magnesium sulphate: with ceramic homogenisers, removes polar organic acids, some sugars and lipids, and carotinoides and chlorophyll, not for use with planar pesticide, 15 ml
Price
273,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Step 2: Dispersive SPE cleanup
Select the dispersive SPE kit suited to the type of food being analysed and the method you are following. In this step, an aliquot of the sample extract from step one (see QuEChERS extraction kits) is added to a 2 or 15 ml centrifuge tube containing a small amount of SPE sorbent and MgSO₄. The sorbent will pull out interfering matrix materials from the sample, while the MgSO₄ helps remove excess water and improve analyte partitioning.
Specificaties
  • Capaciteit:
    15 ml
  • Omschrijving:
    Tube with 400 mg primary secondary amine, 400 mg GCB, 1200 mg magnesium sulphate: with ceramic homogenisers, removes polar organic acids, some sugars and lipids, and carotinoides and chlorophyll, not for use with planar pesticide
  • Seq:
    10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.