Inlaat liners
Liner, ultra inert, 2 mm, dimpled, splitless, 2 mm
Catalogus # HEWL5190-2297
Leverancier: Agilent
Ga naar:
Product Details & Documents
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
- Consistent high level of inertness
Specificaties
- Ø inw.:2 mm
- Omschrijving:Liner, ultra inert, 2 mm, dimpled, splitless
- Seq:11