Orders LinkContactUsLinkComponent
Test gas cans
Test gas cans
Test gas, CH4 - 1.45%, CO2 - 2.5%, CO - 60, H2S - 20, O2 - 15%, capacity 58 l
Catalog # 111-1556
Supplier:   M S A
Test gas cans
Catalog # 111-1556
Supplier:  M S A
Test gas cans
Test gas cans
Test gas, CH4 - 1.45%, CO2 - 2.5%, CO - 60, H2S - 20, O2 - 15%, capacity 58 l
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
These test gases are suitable for use with wearable gas devices.
Specifications
  • Omschrijving:
    Test gas, CH4 - 1.45%, CO2 - 2.5%, CO - 60, H2S - 20, O2 - 15%, capacity 58 l