Test gas cans
Test gas, 50 ppm NH3 in N2, capacity 34 l
Catalogus # 111-1555
Leverancier: M S A
Ga naar:
Product Details & Documents
These test gases are suitable for use with wearable gas devices.
Specificaties
- Omschrijving:Test gas, 50 ppm NH3 in N2, capacity 34 l
- Cat. No.:111-1555