Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Natriumhydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Natriumhydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Poly bottle, 4 L
Catalogus # 3727-03
Corrosief
Irriterend
Leverancier:   Avantor
Natriumhydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Catalogus # 3727-03
Leverancier:  Avantor
CAS-nummer:  1310-73-2
Natriumhydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®
Natriumhydroxide solution 50% (w/w), BAKER ANALYZED®, J.T.Baker®

Poly bottle, 4 L

CAS-nummer:  1310-73-2
Price
316,55 €
Log in voor beschikbaarheid
1 * 4 L
Product Details & Documents

MPORTANT: Material will freeze if stored below 60°F (16°C).

Let op: IMPORTANT: Material will freeze if stored below 60°F (16°C). Protect from air to avoid absorption of carbon dioxide.

Synoniemen:  Caustic soda
  • CAS-nummer:1310-73-2
  • Formule:NaOH
  • MDL-nummer:MFCD00003548
  • Molecular Weight:40 g/mol
  • Kokend Pt:119…135 °C (1013 hPa)
  • Smelten Pt:9…10 °C
  • Dichtheid:1,53 g/cm³ (20 °C)
  • Temperatuur bij opslag:Kamertemperatuur
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II

Documents

Specificaties

Specificatie testresultaten

  • For Laboratory,Research,or Manufacturing Use:
    50 - 52%
  • Protect from air to avoid absorption of carbon dioxide.:
    ≤20
  • Assay (NaOH) (by acidimetry):
    50 - 52 %
  • Color (APHA):
    ≤20
  • Sodium Carbonate (Na₂CO₃):
    ≤0.1 %
  • Chloride (Cl):
    ≤0.002 %
  • Sulfate (SO₄):
    ≤0.001 %
  • Ammonium Hydroxide Precipitate:
    ≤0.01 %
  • Silica (SiO₂), For Information Only:
    ≤5 ppm
  • Heavy Metals (as Ag):
    ≤0.001 %
  • Potassium (K):
    ≤0.01 %
  • Nitrogen Compounds (as N):
    ≤5 ppm
  • Phosphate (PO₄):
    ≤5 ppm
  • Trace Impurities - Iron (Fe):
    ≤5 ppm
  • Trace Impurities - Nickel (Ni):
    ≤5.000 ppm

Specificaties

  • Verpakking:
    Poly bottle
  • VPE:
    4 L

Specificaties

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.