Stopcocks, Galtek®
Inlet 1/4" Flaretek® flared tube, outlet 1/4" Flaretek® flared tube, length 85,9 mm
Catalogus # ENTEPVA4-T4-2FN-1
Leverancier: ENTEGRIS
Ga naar:
Product Details & Documents
Two- or three way, ¹/₈" orifice.
- All wetted parts are PFA and PTFE.
- Small compact design provides minimal internal volume.
- Several connection sizes available to choose from
Specificaties
- Omschrijving:Inlet 1/4" Flaretek® flared tube, outlet 1/4" Flaretek® flared tube, length 85,9 mm