Pass through cabinets
Pass through cabinet, with static dissipative window, sealing gasket and compression handle, 2, 600×400×600 mm
Catalog # PALBPT-604060
Supplier: Palbam
Jump to:
Product Details & Documents
Pass Through cabinet allows products and materials to be transferred between different rooms. Reduces unnecessary walking and entering/exiting cleanroom.
- Single or double versions available
- Sealing gaskets and compression handles ensure firm closure
- Tempered Glass or anti static PVC windows available
- Mechanical or electronic interlocks available
- Can be customised to requirements
Fabrication material: 304 stainless steel with electropolished finish.
Documents
Palbam Class - Catalog 2018
Product Literature
Specifications
- Omschrijving:Pass through cabinet, with static dissipative window, sealing gasket and compression handle
- Seq:01
- b×d×h:600×400×600 mm
- Deuren:2