SNOOD CAP WITH HAIRNET COLOR: DARK BLUE SIZE: M DESCRIPTION: WITH PEAK, LATEXFREE ELASTIC, HEIGHT 8,5CM, DIAMETER OF TOP 19CM MATERIAL: POLYPROPYLENE THICKNESS: PEAK 80G/M, CAP 36G/M, HAIRNET 10-12G/M PACKAGING: 100 PCS/DISPENSERBOX, 1000 PCS/CART 1 * 1.000 ST
Catalogus # GUNA131209
Leverancier: VWR Collection
Frequently Bought Together