Stopcocks, Galtek®
Inlet 1/4" integral ferrule, outlet 1/4" integral ferrule, length 72,9 mm
Catalog # ENTEPVT4-T4-2
Supplier: ENTEGRIS
Jump to:
Product Details & Documents
Two- or three way, ¹/₈" orifice.
- All wetted parts are PFA and PTFE.
- Small compact design provides minimal internal volume.
- Several connection sizes available to choose from
Specifications
- Omschrijving:Inlet 1/4" integral ferrule, outlet 1/4" integral ferrule, length 72,9 mm