Spray gun
Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spool
Catalogus # ENTE421-06
Leverancier: ENTEGRIS
Ga naar:
Product Details & Documents
High purity PFA and PTFE (all wetted parts).
- Orifice: 4,8 mm
- Connection: 1/4" or 1/2" FNPT
- Pressure: max. 5516 mbar
Used in fume hoods and wet processing stations to help protect against corrosion.
Specificaties
- Omschrijving:Repair kit, includes piston, pin-trigger, front and rear diaphragms, spring, spool
- Seq:06



