Spray gun
Replacement cap, blue
Catalogus # ENTE600037BL
Leverancier: ENTEGRIS
Ga naar:
Product Details & Documents
High purity PFA and PTFE (all wetted parts).
- Orifice: 4,8 mm
- Connection: 1/4" or 1/2" FNPT
- Pressure: max. 5516 mbar
Used in fume hoods and wet processing stations to help protect against corrosion.
Specificaties
- Omschrijving:Replacement cap, blue
- Seq:09