Wall-mounted cabinets
Wall-mounted cabinet, with one shelf, lockable, 1 door, left hinged, 600×366×480 mm
Catalogus # KOTT307-00051
Leverancier: KOETTERMANN
Ga naar:
Product Details & Documents
Made of epoxy powder coated steel, with one shelf.
- Solid hinged doors or sliding glass doors
- Height adjustable shelfs
- Maximum loading capacity: 80 kg
Specificaties
- Omschrijving:Wall-mounted cabinet, with one shelf, lockable
- Seq:25
- b×d×h:600×366×480 mm
- Deuren:1 door, left hinged