Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Wall-mounted cabinets

Wall-mounted cabinets

Wall-mounted cabinet, with installation cabinet, 1 door, right hinged, 300×366×780 mm
Catalogus # KOTT307-00036
Leverancier:   KOETTERMANN
Wall-mounted cabinets
Catalogus # KOTT307-00036
Leverancier:  KOETTERMANN
Wall-mounted cabinets
Wall-mounted cabinets

Wall-mounted cabinet, with installation cabinet, 1 door, right hinged, 300×366×780 mm

Price
339,00 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Made of epoxy powder coated steel, with one shelf.

  • Solid hinged doors or sliding glass doors
  • Height adjustable shelfs
  • Maximum loading capacity: 80 kg
Specificaties
  • Omschrijving:
    Wall-mounted cabinet, with installation cabinet
  • Seq:
    05
  • b×d×h:
    300×366×780 mm
  • Deuren:
    1 door, right hinged