Jars with screw cap
150 ml
Catalog # 216-1570
Supplier: Anicrin
Jump to:
Product Details & Documents
Slightly conical vessels, made of PP, translucent with red PE screw cap, for sampling, storage and transportation.
- Twist cap for quick opening and closing
- Very good temperature resistance
- Very good chemical resistance
Caution: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904
Specifications
- Capaciteit:150 ml
- Seq:01