Digestion system, behrotest®
Complete crude fibre digestion unit with 6 sample positions, EXR 6
Catalogus # BEHRB00218448
Leverancier: BEHR
Ga naar:
Product Details & Documents
Digestion apparatus for crude fibre or hydrolysis.
: Includes serial heating unit with individually adjustable heating places, cooling water distribution with condenser rack and hoses, beakers (600 ml), cold fingers.
Specificaties
- Omschrijving:Complete crude fibre digestion unit with 6 sample positions
- Seq:03
- Type:EXR 6