Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Hygromycin B in solution

Hygromycin B in solution

25 mL
Catalogus # A2175.0025
Leverancier:   PanReac AppliChem
Hygromycin B in solution
Catalogus # A2175.0025
Leverancier:  PanReac AppliChem
CAS-nummer:  31282-04-9
Hygromycin B in solution

25 mL

CAS-nummer:  31282-04-9
Price
296,00 €
Log in voor beschikbaarheid
1 * 25 mL
Product Details & Documents
Synoniemen:  (3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
  • CAS-nummer:31282-04-9
  • Formule:C₂₀H₃₇N₃O₁₃
  • MDL-nummer:MFCD06795479
  • Molecular Weight:527.5 g/mol
Specificaties

Specificaties

  • VPE:
    25 mL

Specificaties