Detectable plasters
Detectable plaster strips, blue, refill set for dispenser, 40 pieces, 72×25 mm
Catalogus # SOEH1009984
Leverancier: SOEHNGEN
Ga naar:
Product Details & Documents
These blue, detectable plasters are specially designed for use in the food industry. Dispenser containing aluminium strips with an integrated metal surface that can be detected via a metal detector.
- Plasters are detectable by most metal detectors used within the food industry
- Breathable and skin-friendly
- High quality hypoallergenic plasters
Specificaties
- Omschrijving:Detectable plaster strips, blue, refill set for dispenser, 40 pieces
- l×b:72×25 mm
- Seq:03
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.