Orders LinkContactUsLinkComponent
Detectable plasters
Detectable plasters
Yellow plaster dispenser with 130 blue detectable plasters (plaster strips in 2 sizes, fingertip plasters, finger joint plasters and wound plasters), 160×57×122 mm
Catalog # SOEH1009981
Supplier:   SOEHNGEN
Detectable plasters
Catalog # SOEH1009981
Supplier:  SOEHNGEN
Detectable plasters
Yellow plaster dispenser with 130 blue detectable plasters (plaster strips in 2 sizes, fingertip plasters, finger joint plasters and wound plasters), 160×57×122 mm
Price
Inquire For Price
Log in for availability
1 * 1 SET
Product Details & Documents
These blue, detectable plasters are specially designed for use in the food industry. Dispenser containing aluminium strips with an integrated metal surface that can be detected via a metal detector.
  • Plasters are detectable by most metal detectors used within the food industry
  • Breathable and skin-friendly
  • High quality hypoallergenic plasters
Specifications
  • Omschrijving:
    Yellow plaster dispenser with 130 blue detectable plasters (plaster strips in 2 sizes, fingertip plasters, finger joint plasters and wound plasters)
  • Seq:
    01
  • b×d×h:
    160×57×122 mm