[EN]TUBE 0.5ML V-BOTTOM NON-ST 2D BC RAC 1 * 960 ST
Catalogus # MATRBC83734
Leverancier: THERMO MATRIX TECHNOLOGIES
[EN]TUBE 0.5ML V-BOTTOM NON-ST 2D BC RAC 1 * 960 ST
Catalogus # MATRBC83734
Leverancier: THERMO MATRIX TECHNOLOGIES
Frequently Bought Together
Sealing film for manual application, Parafilm® M
Leverancier: HEATHROW
Aceton ≥99%, TECHNICAL
Leverancier: VWR Chemicals
VWR®, Transferpipetten, Hoge Prestaties
Leverancier: VWR Collection
VWR® NITRILE LIGHT, Nitrilhandschoenen
Leverancier: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Leverancier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Leverancier: VWR Chemicals
VWR®, Serologische pipetten, standaardreeks
Leverancier: VWR Collection
Nitril handschoenen, Kimtech™ Purple Nitrile™
Leverancier: KIMBERLY CLARK
Natriumhydroxide 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur. volumetric solution
Leverancier: VWR Chemicals
pH-teststrips, MQuant®, Supelco®
Leverancier: Merck
VWR®, Nitrilhandschoenen
Leverancier: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Leverancier: VWR Chemicals
VWR®, gezichtstissues
Leverancier: VWR Collection