Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Petrischalen

Petrischalen

Contact plate, with 3 vents, 55 mm, 14 mm, Aseptisch, 25×20/bag
Catalog # ANIC05.20.18PAHW
Supplier:   Anicrin
Petrischalen
Catalog # ANIC05.20.18PAHW
Supplier:  Anicrin
Petrischalen
Petrischalen

Contact plate, with 3 vents, 55 mm, 14 mm, Aseptisch, 25×20/bag

Price
192,00 €
Log in for availability
1 * 500 ST
Product Details & Documents
Specifications
  • Ø:
    55 mm
  • Verpakt:
    25×20/bag
  • Steriliteit:
    Aseptisch
  • Uitvoering:
    Contact plate, with 3 vents
  • Hoogte:
    14 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.