Order Entry
Belgium
ContactUsLinkComponent
MOBCAP COLOR: BLUE SIZE: 52CM DESCRIPTION: WITH DOUBLE LATEXFREE ELASTIC BAND MATERIAL: POLYPROPYLENE THICKNESS: 10-12 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 ST
Catalogus # GUNA128518
Leverancier:  VWR Collection
MOBCAP COLOR: BLUE SIZE: 52CM DESCRIPTION: WITH DOUBLE LATEXFREE ELASTIC BAND MATERIAL: POLYPROPYLENE THICKNESS: 10-12 G/M PACKAGING: 100 PCS/DOUBLE POLYBAG, 1000 PCS/CARTON 1 * 1.000 ST
Catalogus # GUNA128518
Leverancier:  VWR Collection
Frequently Bought Together