Visitor coats, PP
wit, 3XL
Catalogus # 113-9262
Leverancier: VWR Collection
Ga naar:
Product Details & Documents
These disposable coats made of PP are ideal for visitors in non hazardous environments.
- With collar
- Press-stud fastening (5 press-studs)
- No pockets
Weight: 35 g/m²
Colour: White
Documents
Food Contact, BPA, Phthalates Certificate
Aanvullende productdetails
Specificaties
- Confectiemaat:3XL
- Kleur:wit
- Seq:07
- Geslacht:Uniseks
- Cat. No.:113-9262
- Kledingstuk toepassing:General Purpose|Laboratory|Food
- Sluitingstype:Met drukknopen
- Zakken:zonder zakken
- Gewicht:35 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



