Visitor coats, PP
wit, S
Catalogus # 113-9260
Leverancier: VWR Collection
Ga naar:
Product Details & Documents
These disposable coats made of PP are ideal for visitors in non hazardous environments.
- With collar
- Press-stud fastening (5 press-studs)
- No pockets
Weight: 35 g/m²
Colour: White
Documents
Food Contact, BPA, Phthalates Certificate
Aanvullende productdetails
Specificaties
- Confectiemaat:S
- Kleur:wit
- Seq:02
- Geslacht:Uniseks
- Cat. No.:113-9260
- Kledingstuk toepassing:General Purpose|Laboratory|Food
- Sluitingstype:Met drukknopen
- Zakken:zonder zakken
- Gewicht:35 g/m²