Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Visitor coats, PP
Visitor coats, PP
wit, S
Catalogus # 113-9260
Leverancier:   VWR Collection
Visitor coats, PP
Catalogus # 113-9260
Leverancier:  VWR Collection
Visitor coats, PP
Visitor coats, PP
wit, S
Price
58,60 €
Log in voor beschikbaarheid
1 * 50 ST
Product Details & Documents
Specificaties
  • Confectiemaat:
    S
  • Kleur:
    wit
  • Seq:
    02
  • Geslacht:
    Uniseks
  • Cat. No.:
    113-9260
  • Kledingstuk toepassing:
    General Purpose|Laboratory|Food
  • Sluitingstype:
    Met drukknopen
  • Zakken:
    zonder zakken
  • Gewicht:
    35 g/m²
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.