Safety spectacles, Slam
Anti-uv (5-1.4, ESP), anti-scratch, grijs, PC, Smoke
Catalogus # BOLESLAESP
Leverancier: Bollé Safety
Ga naar:
Product Details & Documents
Modern design spectacles with extra-easy fitting and comfort.
- Panoramic visual field
- Anti-scratch, anti-fog, anti-static coatings
- Ultra enveloping
- Comfort temples
Weight: 23 g.
Certifications: EN 166, EN 170 (clear lens), EN 172 (smoke lens).
Delivery information: Supplied with adjustable cord.
Specificaties
- Type:Anti-uv (5-1.4, ESP), anti-scratch
- Kleur:grijs
- Seq:01
- Beglazing:PC, Smoke
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.