Mirrors, Magoptic
Mirror, with focal length of -200 mm, 80 mm
Catalogus # DIDAPOD608417
Leverancier: DIDALAB
Ga naar:
Product Details & Documents
Mirrors used for enabling classroom experiments.
Specificaties
- Omschrijving:Mirror, with focal length of -200 mm
- Seq:11
- Ø:80 mm



