Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Mirrors, Magoptic

Mirrors, Magoptic

Mirror, with focal length of -500 mm, 80 mm
Catalogus # DIDAPOD608481
Leverancier:   DIDALAB
Mirrors, Magoptic
Catalogus # DIDAPOD608481
Leverancier:  DIDALAB
Mirrors, Magoptic
Mirrors, Magoptic

Mirror, with focal length of -500 mm, 80 mm

Price
14,88 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents

Mirrors used for enabling classroom experiments.

Specificaties
  • Omschrijving:
    Mirror, with focal length of -500 mm
  • Seq:
    09
  • Ø:
    80 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.