Orders LinkContactUsLinkComponent
Optical components for optical bench
Optical components for optical bench
Line generator, with a laser, enables to project one line, by the use of one glass cylinder, rotating at 360°
Catalogus # DIDAPOD060220
Leverancier:   DIDALAB
Optical components for optical bench
Catalogus # DIDAPOD060220
Leverancier:  DIDALAB
Optical components for optical bench
Optical components for optical bench
Line generator, with a laser, enables to project one line, by the use of one glass cylinder, rotating at 360°
Price
83,13 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
Specificaties
  • Omschrijving:
    Line generator, with a laser, enables to project one line, by the use of one glass cylinder, rotating at 360°
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Permanent frame, Ø 40 mm
    Permanent frame, Ø 40 mm
    DIDAPOD010095
    Log in voor beschikbaarheid
    1 * 1 ST - 42,87 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Simple lens holder, Ø 40 mm
    Simple lens holder, Ø 40 mm
    DIDAPOD010110
    Log in voor beschikbaarheid
    1 * 1 ST - 21,00 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Lens holder for fragile components, Ø 80 mm
    Lens holder for fragile components, Ø 80 mm
    DIDAPOD010080
    Log in voor beschikbaarheid
    1 * 1 ST - 75,25 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Lens holder, Ø 40/42 mm
    Lens holder, Ø 40/42 mm
    DIDAPOD010090
    Log in voor beschikbaarheid
    1 * 1 ST - 39,38 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Lens holder, Ø 80 mm
    Lens holder, Ø 80 mm
    DIDAPOD060130
    Log in voor beschikbaarheid
    1 * 1 ST - 60,38 €
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mm
    Stand, half moon, for optical components positioned remote from the Optical bench, height 210 mm
    DIDAPOD002192
    Log in voor beschikbaarheid
    1 * 1 ST - 126,00 €