Orders LinkContactUsLinkComponent
Capillary column caps
Capillary column caps
Capillary column caps, silicone
Catalog # REST22858
Supplier:   Restek
Capillary column caps
Catalog # REST22858
Supplier:  Restek
Capillary column caps
Capillary column caps
Capillary column caps, silicone
Price
44,30 €
Log in for availability
1 * 10 ST
Product Details & Documents
These non-reusable column caps increase column lifetime, prevent moisture and air from entering the column during storage.
  • Attach to the column in seconds to form an airtight seal
  • Available in two styles, glass or silicone material
  • Glass caps are colour coded for identifying detector and injector ends
Specifications
  • Omschrijving:
    Capillary column caps, silicone
  • Seq:
    02