Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Polypropylene bottles, Corning®
Wide mouth, Polypropylene, 1000 ml, GL 63
Catalogus # CORN1300P-1LW
Leverancier:   Corning
Polypropylene bottles, Corning®
Catalogus # CORN1300P-1LW
Leverancier:  Corning
Polypropylene bottles, Corning®
Polypropylene bottles, Corning®
Wide mouth, Polypropylene, 1000 ml, GL 63
Price
131,00 €
Log in voor beschikbaarheid
1 * 12 ST
Product Details & Documents
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
  • Hollow glass stopper style
  • Reusable
  • Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.
Documents
Specificaties
  • Schroefdraad:
    GL 63
  • Capaciteit:
    1000 ml
  • Omschrijving:
    Wide mouth, Polypropylene
  • Seq:
    06
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.