Polypropylene bottles, Corning®
Narrow mouth, polypropylene, 250 ml, GL 25
Leverancier: Corning
Ga naar:
Product Details & Documents
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
- Hollow glass stopper style
- Reusable
- Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.
Documents
Certificaten
Zoeken op lotnummerSpecificaties
- Schroefdraad:GL 25
- Capaciteit:250 ml
- Omschrijving:Narrow mouth, polypropylene
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.