Bestelling invoeren
Belgium
Orders LinkContactUsLinkComponent
Polypropylene bottles, Corning®
Polypropylene bottles, Corning®
Wide mouth, Polypropylene, 250 ml, GL 45
Catalogus # CORN1300P-250W
Leverancier:   Corning
Polypropylene bottles, Corning®
Catalogus # CORN1300P-250W
Leverancier:  Corning
Polypropylene bottles, Corning®
Polypropylene bottles, Corning®
Wide mouth, Polypropylene, 250 ml, GL 45
Price
85,20 €
Log in voor beschikbaarheid
1 * 12 ST
Product Details & Documents
Bottles are manufactured from low density polyethylene and are offered with a narrow or wide mouth design.
  • Hollow glass stopper style
  • Reusable
  • Non sterile
The cap and dispensing tube are manufactured from polypropylene to optimize backflow of the solution. The design permits minimal turbulence of the liquid in the bottle.
Documents
Specificaties
  • Schroefdraad:
    GL 45
  • Capaciteit:
    250 ml
  • Omschrijving:
    Wide mouth, Polypropylene
  • Seq:
    04