Orders LinkContactUsLinkComponent
Moppen, MicroMop™
Moppen, MicroMop™
MicroMop® handle, Fibreglass, 1524 mm
Catalogus # MICRMHC-1
Leverancier:   MICRONOVA
Moppen, MicroMop™
Catalogus # MICRMHC-1
Leverancier:  MICRONOVA
Moppen,  MicroMop™
Moppen, MicroMop™
MicroMop® handle, Fibreglass, 1524 mm
Price
63,30 €
Log in voor beschikbaarheid
1 * 1 ST
Product Details & Documents
MicroMop™ are ideal for quick and efficient cleaning of large surface areas. Suitable for wafer processing, electronics, pharmaceutical, and biotech applications. All mop heads attach to stainless steel universal handle via the orange polypropylene quick disconnect adapter. For less critical applications, choose the fibreglass/polypropylene handle with attached adapter.
  • Low-linting, quick drying PVA is ideal for high-tech applications. Can be laundered
  • Polyester has good wicking capability, can be autoclaved and used with strong disinfectants
  • Lite™ is sturdy latex/rayon blend for controlled environments and cleanroom support areas; mop head can be autoclaved for medical and sterile applications
Bestelinformatie: Alle componenten moeten afzonderlijk worden besteld
Specificaties
  • Omschrijving:
    MicroMop® handle, Fibreglass
  • Lengte:
    1524 mm
  • Seq:
    11
Accessories
Productinformatie
  • Catalogus #
    Beschikbaarheid
    Price
  • MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    MicroMop® Adapter: PP clamp adapter to hold Micromop and PolyGen series of floor mops, attaches to SSU series of handles
    129-0170
    Log in voor beschikbaarheid
    1 * 1 ST - 137,00 €